Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: ZFP92 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_005274652.3 | XP_005274709.1 | 6 | 3908 | ||
XM_011531115.2 | XP_011529417.1 | 5 | 3859 | |||
XM_017029281.1 | XP_016884770.1 | 7 | 4257 | |||
GenBank mRNA | AK057819.1 | - | 1 | 224 |