Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAGI2-AS3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038343.2 | 4 | 1006 | |||
GenBank mRNA | AL833456.1 | - | 2 | 730 | ||
CR749529.1 | - | 2 | 1059 |