Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAGI2-AS3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038343.2 | 4 | 8323 | |||
GenBank mRNA | CR749529.1 | - | 3 | 8280 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BX491543.1 | - | 1 | 111 | Chr. 7 - 79467188 - 79467666 [+] on Build GRCh38 |