Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC8A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK095013.1 | - | 1 | 2925 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | DA408888.1 | - | 1 | 355 | Chr. 2 - 40098140 - 40098794 [-] on Build GRCh38 |