Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NAALADL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006713560.2 | XP_006713623.1 | 16 | 8760 | ||
XM_011512612.2 | XP_011510914.1 | 14 | 12014 | |||
XM_011512613.1 | XP_011510915.1 | 14 | 8822 | |||
XM_011512615.2 | XP_011510917.1 | 13 | 11871 | |||
XM_011512616.2 | XP_011510918.1 | 13 | 9552 | |||
XM_017006070.1 | XP_016861559.1 | 18 | 9202 | |||
XM_017006071.1 | XP_016861560.1 | 19 | 9272 | |||
XM_017006072.1 | XP_016861561.1 | 20 | 9406 | |||
XM_017006073.1 | XP_016861562.1 | 18 | 9005 | |||
XM_017006074.1 | XP_016861563.1 | 20 | 11424 | |||
XM_017006075.1 | XP_016861564.1 | 17 | 10149 | |||
XM_017006076.1 | XP_016861565.1 | 17 | 8942 | |||
XM_017006077.1 | XP_016861566.1 | 19 | 10353 | |||
XM_017006078.1 | XP_016861567.1 | 17 | 10170 | |||
XM_017006079.1 | XP_016861568.1 | 15 | 9646 | |||
XM_017006080.1 | XP_016861569.1 | 15 | 10017 | |||
XM_017006081.1 | XP_016861570.1 | 13 | 11867 | |||
XM_017006082.1 | XP_016861571.1 | 15 | 9865 | |||
XM_017006083.1 | XP_016861572.1 | 14 | 8306 | |||
GenBank mRNA | AK021754.1 | - | 1 | 1619 |