Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC105370931 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK124797.1 | - | 1 | 1547 |
Gene Symbol: HDGFRP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006720554.3 | XP_006720617.1 | 6 | 3205 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BM931913.1 | - | 1 | 152 | Chr. 15 - 83112750 - 83113434 [-] on Build GRCh38 |