Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GPATCH2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018040.3 | NP_060510.1 | 10 | 5288 | ||
GenBank mRNA | AK123745.1 | - | 1 | 1293 |