Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HIP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AY358103.1 | - | 1 | 298 | ||
JX965370.1 | - | 2 | 466 | |||
JX965371.1 | - | 2 | 348 |