Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZBTB7C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039360.2 | NP_001034449.1 | 3 | 4088 | ||
NM_001318841.1 | NP_001305770.1 | 5 | 4269 | |||
XM_005258229.4 | XP_005258286.1 | 4 | 4013 | |||
XM_011525861.2 | XP_011524163.1 | 7 | 4493 | |||
XM_011525863.2 | XP_011524165.1 | 6 | 4610 | |||
XM_011525864.2 | XP_011524166.1 | 6 | 4365 | |||
XM_011525865.2 | XP_011524167.1 | 6 | 4606 | |||
XM_011525866.2 | XP_011524168.1 | 5 | 4351 | |||
XM_011525867.2 | XP_011524169.1 | 5 | 4140 | |||
XM_011525869.2 | XP_011524171.1 | 6 | 4615 | |||
XM_011525870.2 | XP_011524172.1 | 5 | 4311 | |||
XM_011525871.2 | XP_011524173.1 | 3 | 4826 | |||
XM_017025605.1 | XP_016881094.1 | 7 | 6288 | |||
XM_017025606.1 | XP_016881095.1 | 7 | 4742 | |||
XM_017025607.1 | XP_016881096.1 | 7 | 4497 | |||
XM_017025608.1 | XP_016881097.1 | 6 | 6165 | |||
XM_017025609.1 | XP_016881098.1 | 5 | 4482 | |||
GenBank mRNA | AK095226.1 | - | 1 | 1825 | ||
AM988836.1 | - | 3 | 4076 | |||
AM988837.1 | - | 3 | 4221 | |||
AM988838.1 | - | 4 | 4271 | |||
AM988839.1 | - | 5 | 4269 |