Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZBTB7C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_005258229.4 | XP_005258286.1 | 4 | 4293 | ||
XM_011525861.2 | XP_011524163.1 | 7 | 4773 | |||
XM_011525863.2 | XP_011524165.1 | 6 | 4890 | |||
XM_011525864.2 | XP_011524166.1 | 6 | 4645 | |||
XM_011525865.2 | XP_011524167.1 | 6 | 4886 | |||
XM_011525866.2 | XP_011524168.1 | 5 | 4631 | |||
XM_011525867.2 | XP_011524169.1 | 5 | 4420 | |||
XM_011525869.2 | XP_011524171.1 | 6 | 4895 | |||
XM_011525870.2 | XP_011524172.1 | 5 | 4591 | |||
XM_011525871.2 | XP_011524173.1 | 3 | 5106 | |||
XM_017025605.1 | XP_016881094.1 | 7 | 6568 | |||
XM_017025606.1 | XP_016881095.1 | 7 | 5022 | |||
XM_017025607.1 | XP_016881096.1 | 7 | 4777 | |||
XM_017025608.1 | XP_016881097.1 | 6 | 6445 | |||
XM_017025609.1 | XP_016881098.1 | 5 | 4762 | |||
GenBank mRNA | AK095226.1 | - | 2 | 1927 |