Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other NRG1 SIRNA Assays ›
Gene Symbol:
NRG1
Gene Name:
neuregulin 1
Gene Aliases:
ARIA, GGF, GGF2, HGL, HRG, HRG1, HRGA, MST131, MSTP131, NDF, NRG1-IT2, SMDF
Chromosome Location:
Chr.8: 31639222 - 32771716 on Build GRCh38
UniGene ID:
Hs.453951
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
n364073
Purification
Standard
HPLC
HPLC-IVR
Size
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
Availability
Made To Order
Catalog #
4390771
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: NRG1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001159995.2
NP_001153467.1
12
5251
NM_001159996.2
NP_001153468.1
9
5107
NM_001159999.2
NP_001153471.1
13
5302
NM_001160001.2
NP_001153473.1
11
5200
NM_001160004.2
NP_001153476.1
13
5827
NM_001322197.1
NP_001309126.1
8
4956
NM_001322201.1
NP_001309130.1
10
5168
NM_001322202.1
NP_001309131.1
9
5117
NM_001322205.1
NP_001309134.1
9
6343
NM_001322206.1
NP_001309135.1
10
6485
NM_013956.4
NP_039250.2
13
5709
NM_013957.4
NP_039251.2
12
5685
NM_013960.4
NP_039254.1
13
5836
NM_013964.4
NP_039258.1
12
5694
XM_005273486.3
XP_005273543.1
8
6328
XM_005273487.3
XP_005273544.1
8
6319
XM_006716335.3
XP_006716398.1
9
4694
XM_011544512.2
XP_011542814.1
13
5492
XM_017013365.1
XP_016868854.1
12
5406
XM_017013366.1
XP_016868855.1
12
5397
XM_017013367.1
XP_016868856.1
11
5202
XM_017013368.1
XP_016868857.1
11
5539
XM_017013369.1
XP_016868858.1
9
6311
XM_017013370.1
XP_016868859.1
8
6291
GenBank mRNA
AF176921.1
-
1
1532
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
Transcript Location
GenBankmRNA
BM553545.1
-
695
Chr. - - [-] on Build
Back To Top
More Information
Panther Classification:
Molecular Function -
growth factor
Gene Ontology Categories:
Function(s)
Process(es)
MAPK cascade
activation of MAPK activity
cell morphogenesis
startle response
cardiac conduction system development
ventricular trabecula myocardium morphogenesis
cell communication
activation of transmembrane receptor protein tyrosine kinase activity
nervous system development
synapse assembly
peripheral nervous system development
locomotory behavior
regulation of blood pressure
positive regulation of cell proliferation
embryo development
neural crest cell development
regulation of phosphatidylinositol 3-kinase signaling
positive regulation of phosphatidylinositol 3-kinase signaling
glucose transport
peptidyl-tyrosine phosphorylation
glial cell fate commitment
chemorepulsion involved in interneuron migration from the subpallium to the cortex
mammary gland development
positive regulation of myelination
activation of protein kinase B activity
intracellular signal transduction
ERBB signaling pathway
ERBB2 signaling pathway
wound healing
negative regulation of protein catabolic process
myelination
regulation of protein homodimerization activity
regulation of protein heterodimerization activity
positive regulation of GTPase activity
neurotransmitter receptor metabolic process
positive regulation of axon extension
negative regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
positive regulation of Ras protein signal transduction
phosphatidylinositol phosphorylation
phosphatidylinositol-mediated signaling
neuron fate commitment
oligodendrocyte differentiation
positive regulation of peptidyl-tyrosine phosphorylation
negative regulation of secretion
positive regulation of striated muscle cell differentiation
positive regulation of protein kinase B signaling
cardiac muscle cell differentiation
cardiac muscle cell myoblast differentiation
endocardial cell differentiation
positive regulation of dendritic spine development
positive regulation of calcineurin-NFAT signaling cascade
positive regulation of protein targeting to mitochondrion
positive regulation of protein localization to cell surface
regulation of cell motility
positive regulation of cardiac muscle cell differentiation
negative regulation of neuron migration
transcription cofactor activity
protein tyrosine kinase activity
Ras guanyl-nucleotide exchange factor activity
cytokine activity
ErbB-2 class receptor binding
integrin binding
growth factor activity
transmembrane receptor protein tyrosine kinase activator activity
receptor tyrosine kinase binding
ErbB-3 class receptor binding
chemorepellent activity
phosphatidylinositol-4,5-bisphosphate 3-kinase activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5d97d974fc-8gqds:80/10.239.164.183:80.
git-commit: fe8b04579249ff98075bad6061a6e2a4739e01ce
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.13.0-2024.08.32-1.0