Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FLRT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_005267490.3 | XP_005267547.1 | 2 | 7803 | ||
XM_017021129.1 | XP_016876618.1 | 2 | 7985 | |||
XM_017021130.1 | XP_016876619.1 | 2 | 17259 | |||
XM_017021131.1 | XP_016876620.1 | 3 | 16413 | |||
XM_017021132.1 | XP_016876621.1 | 3 | 7892 | |||
XM_017021133.1 | XP_016876622.1 | 3 | 7910 | |||
XM_017021134.1 | XP_016876623.1 | 3 | 7837 |