Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM98B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173611.3 | NP_775882.2 | 8 | 3731 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | CR607099.1 | - | 1 | 1646 | Chr. 15 - 38485408 - 38487080 [+] on Build GRCh38 |