Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FBXO32 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001242463.1 | NP_001229392.1 | 7 | 2435 | ||
NM_058229.3 | NP_478136.1 | 9 | 2714 | |||
NM_148177.2 | NP_680482.1 | 6 | 2349 | |||
GenBank mRNA | AK125888.1 | - | 1 | 853 |