Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CSNK1D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_005256337.4 | XP_005256394.1 | 9 | 2113 |
Gene Symbol: SLC16A3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BM980920.1 | - | 1 | 37 |