Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DIS3L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_005254146.3 | XP_005254203.1 | 15 | 3492 | ||
GenBank mRNA | AF147431.1 | - | 1 | 491 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | CD358046.1 | - | 233 | Chr. - - [-] on Build |