Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FUT9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC001879.1 | - | 3 | 502 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BE266935.1 | - | 3 | 505 | Chr. 6 - 96015984 - 96036888 [+] on Build GRCh38 |