Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: LOC283737 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK096897.1 | - | 1 | 171 |
Gene Symbol: AGBL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011521226.2 | XP_011519528.1 | 23 | 11023 | ||
XM_017021918.1 | XP_016877407.1 | 23 | 10990 | |||
XM_017021919.1 | XP_016877408.1 | 22 | 10939 |