Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LINC00921 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033904.1 | 1 | 118 | |||
GenBank mRNA | AK096958.1 | - | 1 | 84 | ||
BC104941.1 | - | 1 | 143 | |||
BC104943.1 | - | 1 | 143 | |||
DA532447.1 | - | 1 | 120 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000573951.1 | - | 1 | 143 | Chr. 16 - 3263743 - 3267567 [+] on Build GRCh38 |