Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IGHM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC001872.1 | - | 2 | 117 | ||
BC002963.1 | - | 2 | 97 | |||
BC006180.1 | - | 2 | 117 | |||
BC017356.2 | - | 2 | 114 | |||
BC019235.2 | - | 2 | 116 | |||
BC073767.1 | - | 2 | 105 |
Gene Symbol: SKAP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF174049.1 | - | 1 | 33 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BC013877.1 | - | 987 | Chr. - - [-] on Build |