Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IGHM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC001872.1 | - | 2 | 301 | ||
BC002963.1 | - | 2 | 281 | |||
BC006180.1 | - | 2 | 301 | |||
BC017356.2 | - | 2 | 298 | |||
BC019235.2 | - | 2 | 300 | |||
BC073767.1 | - | 2 | 289 |
Gene Symbol: LOC102723407 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011546198.1 | XP_011544500.1 | 2 | 339 | ||
GenBank mRNA | AF062240.1 | - | 2 | 277 | ||
AF103048.1 | - | 1 | 195 | |||
AF103313.1 | - | 1 | 187 | |||
AF174124.1 | - | 220 | ||||
AF396470.1 | - | 1 | 220 | |||
AF396478.1 | - | 1 | 220 | |||
AF455548.1 | - | 1 | 220 | |||
AJ244933.1 | - | 1 | 220 | |||
AJ244948.1 | - | 1 | 220 | |||
AJ244980.1 | - | 1 | 220 | |||
AJ389176.1 | - | 1 | 220 | |||
AJ414019.1 | - | 1 | 220 | |||
AJ579202.1 | - | 1, 4 | 223 | |||
AY376082.1 | - | 1 | 220 | |||
AY393167.1 | - | 1 | 173 | |||
AY671339.1 | - | 1 | 213 | |||
DQ100904.1 | - | 1 | 216 | |||
DQ454522.1 | - | 1 | 142 | |||
DQ454827.1 | - | 1 | 142 | |||
DQ454834.1 | - | 1 | 136 | |||
DQ454943.1 | - | 1 | 139 | |||
DQ454953.1 | - | 1 | 139 | |||
DQ523972.1 | - | 217 | ||||
DQ523999.1 | - | 1 | 217 | |||
DQ524000.1 | - | 1 | 217 | |||
EF175423.1 | - | 155 | ||||
EF175437.1 | - | 155 | ||||
EU099126.1 | - | 1 | 169 | |||
FJ039783.1 | - | 1 | 220 | |||
HM152512.1 | - | 1 | 220 | |||
HM152518.1 | - | 2 | 223 | |||
JN376176.1 | - | 1 | 145 | |||
JN376188.1 | - | 1 | 145 | |||
JN376230.1 | - | 1 | 145 | |||
JN376231.1 | - | 1 | 145 | |||
JN376232.1 | - | 1 | 145 | |||
JN376233.1 | - | 1 | 145 | |||
JN376234.1 | - | 1 | 145 | |||
JN376235.1 | - | 1 | 145 | |||
JN376309.1 | - | 1 | 145 | |||
KU674890.1 | - | 1 | 173 | |||
S45207.1 | - | 385 | ||||
S45947.1 | - | 277 | ||||
Y14936.1 | - | 1 | 199 | |||
Z14195.1 | - | 2 | 314 | |||
Z14206.1 | - | 2 | 313 |
Gene Symbol: SKAP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF174049.1 | - | 1 | 217 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BC013877.1 | - | 1171 | Chr. - - [-] on Build |