Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PCOLCE-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038910.1 | 7 | 1674 | |||
GenBank mRNA | AK097705.1 | - | 7 | 1674 | ||
AK298216.1 | - | 8 | 1142 |