Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DYNC1I2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271785.1 | NP_001258714.1 | 18 | 3491 | ||
NM_001271786.1 | NP_001258715.1 | 16 | 3484 | |||
NM_001271787.1 | NP_001258716.1 | 16 | 3467 | |||
NM_001271788.1 | NP_001258717.1 | 16 | 3430 | |||
NM_001271789.1 | NP_001258718.1 | 16 | 3413 | |||
NM_001271790.1 | NP_001258719.1 | 16 | 3427 | |||
NM_001320882.1 | NP_001307811.1 | 17 | 3490 | |||
NM_001320883.1 | NP_001307812.1 | 17 | 3473 | |||
NM_001320884.1 | NP_001307813.1 | 16 | 3410 | |||
NM_001378.2 | NP_001369.1 | 18 | 3508 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | CR601532.1 | - | 1 | 1446 | Chr. 2 - 171747807 - 171749697 [+] on Build GRCh38 |