Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRAF3IP2-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_034108.1 | 4 | 4850 | |||
NR_034109.1 | 2 | 4559 | ||||
GenBank mRNA | AF136408.1 | - | 1 | 2328 |