Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Osr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK017286.1 | - | 2 | 750 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY717095.1 | - | 2 | 748 | Chr. 15 - 35296098 - 35297409 [+] on Build GRCm38 |