Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gm20740 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045279.1 | 3, 4 | 282 | |||
GenBank mRNA | AK040104.1 | - | 3, 4 | 283 |