Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Skint1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK029209.1 | - | 2 | 1531 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AV243596.2 | - | 1 | 497 | Chr. 4 - 112011645 - 112012343 [+] on Build GRCm38 |