Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Taf6l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK013112.1 | - | 1 | 389 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BU758277.1 | - | 1 | 337 | Chr. 19 - 8780462 - 8781001 [-] on Build GRCm38 |