Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arhgap15os | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040622.1 | 3 | 1333 | |||
GenBank mRNA | AK078625.1 | - | 3 | 1332 |