Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arhgap15os | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040622.1 | 1 | 37 | |||
GenBank mRNA | AK078625.1 | - | 1 | 38 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY719583.1 | - | 1 | 38 | Chr. 2 - 44060352 - 44065324 [-] on Build GRCm38 |