Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 1700037J18Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK006623.1 | - | 2 | 136 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY706620.1 | - | 2 | 135 | Chr. 9 - 16599380 - 16599802 [+] on Build GRCm38 |