Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 4933406B15Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK016679.1 | - | 2 | 536 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY716368.1 | - | 536 | Chr. - - [-] on Build |