Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prox2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK086375.1 | - | 1, 2 | 365 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB662944.1 | - | 1, 2 | 365 | Chr. 12 - 85086374 - 85087707 [+] on Build GRCm38 |