Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Junos | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK076859.1 | - | 2 | 152 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB615473.1 | - | 2 | 152 | Chr. 4 - 95123995 - 95165883 [+] on Build GRCm38 |