Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ar | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK037070.1 | - | 1843 | |||
AK054123.1 | - | 1959 | ||||
AK162901.1 | - | 1887 |