Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gm15133 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040749.1 | 7 | 803 | |||
GenBank mRNA | AK085986.1 | - | 7 | 804 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB519837.2 | - | 5 | 623 | Chr. 7 - 104317642 - 104324825 [+] on Build GRCm38 |