Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 2900022M12Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK035467.1 | - | 1 | 2837 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB115885.2 | - | 1 | 475 | Chr. 16 - 92684225 - 92684804 [-] on Build GRCm38 |