Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gm29687 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK139823.1 | - | 3 | 328 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY725917.1 | - | 3 | 328 | Chr. 12 - 26092182 - 26210505 [-] on Build GRCm38 |