Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 9130015A21Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045050.1 | 3 | 417 | |||
GenBank mRNA | AK018624.1 | - | 3 | 417 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY720805.1 | - | 3 | 413 | Chr. 12 - 35672388 - 35695114 [-] on Build GRCm38 |