Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: 2610100L16Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
RefSeq | NR_033490.1 | 3 | 727 | |||
GenBank mRNA | AK011787.1 | - | LIT3367 2610100L16 |
3 | 702 | |
AK163433.1 | - | 3 | 600 |