Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 1110014L15Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK003465.1 | - | 1 | 774 | ||
AK003710.1 | - | 1 | 996 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY703724.1 | - | 1 | 774 | Chr. 7 - 6671741 - 6672670 [-] on Build GRCm38 |