Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 4930414N08Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK015140.1 | - | 1 | 298 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY714688.1 | - | 298 | Chr. - - [-] on Build |