Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Limk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK042206.1 | - | 1 | 523 | ||
AK044582.1 | - | 1 | 526 | |||
AK161560.1 | - | 1 | 513 |