Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dnah2os | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK020071.1 | - | 2 | 190 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY718144.1 | - | 2 | 188 | Chr. 11 - 69448548 - 69453369 [+] on Build GRCm38 |