Search Thermo Fisher Scientific
Gene Symbol: Gm31087 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC049679.1 | - | 177 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BU848365.1 | - | 1 | 180 | Chr. 18 - 40242753 - 40258495 [-] on Build GRCm38 |