- Order Status
- Quick Order
-
Don't have an account ? Create Account
Gene Symbol: 1500017E21Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033510.1 | 5 | 732 | |||
GenBank mRNA | AK149390.1 | - | 4 | 363 | ||
AK149600.1 | - | 3 | 315 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | CJ133907.1 | - | 4 | 363 | Chr. 19 - 36621033 - 36626024 [-] on Build GRCm38 |