Gene Symbol: 1500017E21Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033510.1 | 5 | 1296 | |||
GenBank mRNA | AK142254.1 | - | 5 | 1300 | ||
AK149390.1 | - | 4 | 927 | |||
AK149600.1 | - | 3 | 879 |
Gene Symbol: Hectd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BP754340.1 | - | 1 | 370 |