Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 2900022M12Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK013577.1 | - | 1 | 533 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BF100716.1 | - | 36 | Chr. - - [-] on Build |