Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gm16087 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK132125.1 | - | 1 | 435 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY713232.1 | - | 1 | 431 | Chr. 12 - 106954767 - 106955924 [-] on Build GRCm38 |