Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 1700061D14Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK006850.1 | - | 2 | 306 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY706831.1 | - | 2 | 306 | Chr. 5 - 37969955 - 37983601 [-] on Build GRCm38 |